Assay Details
| Gene Symbol: C22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182520.2 | NP_872326.2 | 3 - 4 | 536 | 65 | |
| XM_011529907.2 | XP_011528209.1 | 2 - 3 | 1327 | 65 | ||
| XM_017028612.1 | XP_016884101.1 | 3 - 4 | 1032 | 65 | ||
| XM_017028614.1 | XP_016884103.1 | 2 - 3 | 1327 | 65 | ||
| GenBank mRNA | BU739625.1 | - | 3 - 4 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000137.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | ENST00000305199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | ENST00000336186.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | ENST00000402217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002392.hg.1 | TC22000137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | ENST00000305199.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | ENST00000336186.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | ENST00000402217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002397.hg.1 | TC22000137.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | TC2200006860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147276.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | ENST00000498542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006860.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | NM_182520.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | TC2200006860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000305199.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000382821.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000402217.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | ENST00000477921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147271.hg.1 | NM_182520.2 | Refseq |

C22orf15
Genomic Map