Assay Details
| Gene Symbol: ARID3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259513.4 | XP_005259570.1 | 1 - 2 | 479 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259513.4 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
protein binding
protein homodimerization activity
transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000592216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | ENST00000592216.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000263620.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000457152.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585733.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000585895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000587532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000590749.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000592216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | ENST00000592216.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006499.hg.1 | NM_005224.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009120.hg.1 | ENST00000585647.1 | Ensembl |

ARID3A
Genomic Map