Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004454.2 | NP_004445.1 | 4 - 5 | 429 | 120 | |
| GenBank mRNA | AK301878.1 | - | 4 - 5 | 344 | 120 | |
| BC007333.2 | - | 4 - 5 | 392 | 3050350 | 120 | |
| BT006713.1 | - | 3 - 4 | 182 | 120 | ||
| CR456767.1 | - | 3 - 4 | 182 | 120 | ||
| X76184.1 | - | 4 - 5 | 377 | 120 | ||
More Information
Set Membership: |
|

ETV5
Genomic Map