Assay Details
| Gene Symbol: ETFBKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173802.3 | NP_776163.1 | 1 - 2 | 178 | 63 | |
| GenBank mRNA | BC039535.1 | - | 1 - 2 | 178 | 5744381 | 63 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000293.hg
Additional Information:
This assay is in the 5'UTR region of the BC039535.1,NM_173802.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | ENST00000395763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | ENST00000412352.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | ENST00000538463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003475.hg.1 | TC12000293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003468.hg.1 | ENST00000395763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003468.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003468.hg.1 | TC12000293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | ENST00000395763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | ENST00000412352.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | ENST00000538463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003476.hg.1 | TC12000293.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150827.hg.1 | TC1200007236.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007236.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000357721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000412352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000538463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | ENST00000539633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_001135863.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_001135864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150825.hg.1 | TC1200007236.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200150818.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150818.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150818.hg.1 | ENST00000395763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150818.hg.1 | NM_173802.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150818.hg.1 | TC1200007236.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 183413 | - | - |

ETFBKMT
Genomic Map