Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
ENST00000348476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
ENST00000355431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
ENST00000395168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
ENST00000431317.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
NM_002892.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
NM_023000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
NM_023001.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003753.hg.1 |
TC14000348.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000348476.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000355431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000395168.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000431317.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000466065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_002892.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_023000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_023001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000348476.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000355431.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000395168.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000431317.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
ENST00000466065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_002892.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_023000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
NM_023001.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400150695.hg.1 |
TC1400007307.hg.1 |
Clariom D TC |