Assay Details
| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018011.3 | NP_060481.3 | 1 - 2 | 597 | 82 | |
| GenBank mRNA | AK001016.1 | - | 1 - 2 | 595 | 82 | |
| AK303575.1 | - | 2 - 3 | 370 | 82 | ||
| AK304751.1 | - | 2 - 3 | 427 | 82 | ||
| BC050434.1 | - | 1 - 2 | 544 | 6016552 | 82 | |
| BC071587.1 | - | 1 - 2 | 568 | 5271043 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000861.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13009650.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009650.hg.1 | ENST00000426600.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009650.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009650.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009652.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009652.hg.1 | ENST00000426600.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009652.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009652.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007943.hg.1 | ENST00000610791.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009649.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009649.hg.1 | ENST00000426600.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009649.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009649.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009651.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009651.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009651.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009645.hg.1 | ENST00000375926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009645.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009645.hg.1 | ENST00000426600.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009645.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009645.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300007943.hg.1 | ENST00000610791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164908.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300164912.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164912.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164912.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164912.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164912.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164910.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164910.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164910.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164910.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164910.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164905.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300164909.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164909.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164909.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164909.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164909.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300164904.hg.1 | TC1300009714.hg.1 | Clariom D TC |

ARGLU1
Genomic Map