Assay Details
| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018011.3 | NP_060481.3 | 2 - 3 | 816 | 79 | |
| GenBank mRNA | AK001016.1 | - | 2 - 3 | 814 | 79 | |
| BC050434.1 | - | 2 - 3 | 763 | 6016552 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000861.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13009638.hg.1 | ENST00000375926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009638.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009638.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009638.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009638.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009646.hg.1 | ENST00000375926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009646.hg.1 | ENST00000400198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009646.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009646.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009646.hg.1 | TC13000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164899.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164900.hg.1 | TC1300009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000360629.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000375926.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000400198.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | ENST00000472226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009714.hg.1 | NM_018011.3 | Refseq |

ARGLU1
Genomic Map