Assay Details
| Gene Symbol: ARHGDIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321420.1 | NP_001308349.1 | 2 - 3 | 390 | 158 | |
| GenBank mRNA | BG390701.1 | - | 2 - 3 | 207 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001321420.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
negative regulation of cell adhesion
multicellular organism development
regulation of Rho protein signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cellular response to redox state
negative regulation of trophoblast cell migration
regulation of actin cytoskeleton reorganization
GTPase activity
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Rac GTPase binding
movement of cell or subcellular component
negative regulation of cell adhesion
multicellular organism development
regulation of Rho protein signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cellular response to redox state
negative regulation of trophoblast cell migration
regulation of actin cytoskeleton reorganization
GTPase activity
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Rac GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000228945.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000535676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000536592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000539131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000541644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000542276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | ENST00000545895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010024.hg.1 | NM_001175.6 | Refseq |

ARHGDIB
Genomic Map