Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK292051.1 | - | 1 - 2 | 102 | 73 | |
| BC014315.1 | - | 1 - 2 | 122 | 3997264 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010883.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184992.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184993.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184995.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000395589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000562411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000563480.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000566658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184999.hg.1 | NM_006049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000568875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | TC1500010883.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000307979.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000316634.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000565465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184991.hg.1 | ENST00000568875.1 | Ensembl |

SNAPC5
Genomic Map