| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
ENST00000262367.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
ENST00000323508.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
ENST00000382070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
ENST00000543883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
NM_001079846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
NM_004380.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011937.hg.1 |
TC16000823.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
ENST00000262367.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
ENST00000382070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
NM_001079846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
NM_004380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
TC1600009202.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
NM_004380.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
ENST00000262367.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
ENST00000382070.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170361.hg.1 |
NM_001079846.1 |
Refseq |