Assay Details
| Gene Symbol: TMEM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526853.2 | XP_011525155.1 | 1 - 2 | 143 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011526853.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000326652.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000585918.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000586609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000591570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | ENST00000592891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001085488.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NM_001282011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104066.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011464.hg.1 | NR_104071.1 | Refseq |

TMEM150B
Genomic Map