| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025017.hg.1 |
ENST00000261402.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025017.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025017.hg.1 |
TC12001916.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025018.hg.1 |
ENST00000261402.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025018.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025018.hg.1 |
TC12001916.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200191398.hg.1 |
TC1200011786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200191399.hg.1 |
TC1200011786.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000261402.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000548902.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000549704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
ENST00000553094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011786.hg.1 |
NM_014840.2 |
Refseq |