Assay Details
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL713727.1 | - | 8 - 9 | 1735 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
