| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022681.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022681.hg.1 |
ENST00000541232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022681.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022681.hg.1 |
TC04001625.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012068.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000322396.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000510269.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000511687.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
ENST00000514886.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
NM_001304458.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
NM_138364.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182877.hg.1 |
TC0400012068.hg.1 |
Clariom D TC |