Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304458.1 | NP_001291387.1 | 1 - 2 | 435 | 68 | |
| XM_017008825.1 | XP_016864314.1 | 1 - 2 | 346 | 68 | ||
| XM_017008826.1 | XP_016864315.1 | 1 - 2 | 309 | 68 | ||
| GenBank mRNA | AK096703.1 | - | 1 - 2 | 330 | 68 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK096703.1,NM_001304458.1,XM_017008825.1,XM_017008826.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182888.hg.1 | TC0400012068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |

PRMT9
Genomic Map