Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304458.1 | NP_001291387.1 | 6 - 7 | 1238 | 149 | |
| NM_138364.3 | NP_612373.2 | 7 - 8 | 1387 | 149 | ||
| XM_017008823.1 | XP_016864312.1 | 7 - 8 | 1128 | 149 | ||
| XM_017008824.1 | XP_016864313.1 | 6 - 7 | 1060 | 149 | ||
| XM_017008825.1 | XP_016864314.1 | 6 - 7 | 1114 | 149 | ||
| XM_017008826.1 | XP_016864315.1 | 5 - 6 | 1009 | 149 | ||
| GenBank mRNA | AK096703.1 | - | 6 - 7 | 1133 | 149 | |
| AK128483.1 | - | 1 - 2 | 470 | 149 | ||
| AK292904.1 | - | 7 - 8 | 1320 | 149 | ||
| BC021250.2 | - | 7 - 8 | 1142 | 149 | ||
| BC064403.1 | - | 7 - 8 | 1286 | 5547611 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182882.hg.1 | TC0400012068.hg.1 | Clariom D TC |

PRMT9
Genomic Map