Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304458.1 | NP_001291387.1 | 7 - 8 | 1418 | 119 | |
| NM_138364.3 | NP_612373.2 | 8 - 9 | 1567 | 119 | ||
| XM_017008823.1 | XP_016864312.1 | 8 - 9 | 1308 | 119 | ||
| XM_017008824.1 | XP_016864313.1 | 7 - 8 | 1240 | 119 | ||
| XM_017008825.1 | XP_016864314.1 | 7 - 8 | 1294 | 119 | ||
| XM_017008826.1 | XP_016864315.1 | 6 - 7 | 1189 | 119 | ||
| GenBank mRNA | AK096703.1 | - | 7 - 8 | 1313 | 119 | |
| AK128483.1 | - | 2 - 3 | 650 | 119 | ||
| AK292904.1 | - | 8 - 9 | 1500 | 119 | ||
| BC021250.2 | - | 8 - 9 | 1322 | 119 | ||
| BC064403.1 | - | 8 - 9 | 1466 | 5547611 | 119 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001625.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022685.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022685.hg.1 | ENST00000541232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022685.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022685.hg.1 | TC04001625.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182881.hg.1 | TC0400012068.hg.1 | Clariom D TC |

PRMT9
Genomic Map