| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000591.hg.1 |
ENST00000179259.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000591.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000591.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000591.hg.1 |
TC12000061.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000539671.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
ENST00000539671.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000539671.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
ENST00000539671.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006557.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
ENST00000179259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
ENST00000537251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
NM_020375.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200145092.hg.1 |
TC1200006557.hg.1 |
Clariom D TC |