Assay Details
| Gene Symbol: LAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012261.3 | NP_036393.1 | 3 - 4 | 869 | 106 | |
| GenBank mRNA | AL121740.1 | - | 3 - 4 | 576 | 106 | |
| BC050727.1 | - | 3 - 4 | 572 | 6044324 | 106 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000077.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001136.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001136.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001136.hg.1 | TC20000077.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001137.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001137.hg.1 | ENST00000427562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001137.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001137.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001137.hg.1 | TC20000077.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006676.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | TC2000006676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | ENST00000427562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | NM_001199897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145804.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145803.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145803.hg.1 | NM_012261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000145803.hg.1 | TC2000006676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000145803.hg.1 | ENST00000246070.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000145803.hg.1 | NM_012261.3 | Refseq |

LAMP5
Genomic Map