Assay Details
| Gene Symbol: VRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003384.2 | NP_003375.1 | 11 - 12 | 1175 | 77 | |
| XM_006720247.3 | XP_006720310.1 | 11 - 12 | 1213 | 77 | ||
| XM_017021622.1 | XP_016877111.1 | 10 - 11 | 1074 | 77 | ||
| XM_017021625.1 | XP_016877114.1 | 11 - 12 | 1254 | 77 | ||
| GenBank mRNA | AB000449.1 | - | 11 - 12 | 1144 | 77 | |
| AK290110.1 | - | 11 - 12 | 1163 | 77 | ||
| BC103761.1 | - | 11 - 12 | 1153 | 5773877 | 77 | |
| BC112075.1 | - | 11 - 12 | 1143 | 8327543 | 77 | |
| BC113510.1 | - | 11 - 12 | 1143 | 8322562 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000608.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
mitotic nuclear division
mitotic nuclear envelope disassembly
mitotic nuclear envelope reassembly
regulation of cell shape
histone H3-S10 phosphorylation
protein autophosphorylation
cell division
histone H3-T3 phosphorylation
Golgi disassembly
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)
protein phosphorylation
mitotic nuclear division
mitotic nuclear envelope disassembly
mitotic nuclear envelope reassembly
regulation of cell shape
histone H3-S10 phosphorylation
protein autophosphorylation
cell division
histone H3-T3 phosphorylation
Golgi disassembly
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14007581.hg.1 | ENST00000216639.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007581.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007581.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007581.hg.1 | TC14000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007583.hg.1 | ENST00000216639.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007583.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007583.hg.1 | TC14000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007584.hg.1 | ENST00000216639.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007584.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007584.hg.1 | TC14000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007582.hg.1 | ENST00000216639.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007582.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007582.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007582.hg.1 | TC14000608.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180544.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180545.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180548.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180548.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180548.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180548.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180548.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180549.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180549.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180549.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180549.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180549.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180546.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180541.hg.1 | TC1400010649.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000216639.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000553683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000555402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | ENST00000557352.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010649.hg.1 | NM_003384.2 | Refseq |

VRK1
Genomic Map