| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005573.hg.1 |
ENST00000360454.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005573.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005573.hg.1 |
ENST00000481904.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005573.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005573.hg.1 |
TC06000520.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005570.hg.1 |
ENST00000360454.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005570.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005570.hg.1 |
ENST00000481904.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005570.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005570.hg.1 |
TC06000520.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600154140.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154140.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154140.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154140.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154140.hg.1 |
TC0600007819.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000467122.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007819.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000360454.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000403376.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
ENST00000481904.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NM_001286550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NM_207409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NR_104467.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
NR_104469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154136.hg.1 |
TC0600007819.hg.1 |
Clariom D TC |