Assay Details
| Gene Symbol: ZDHHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185024.1 | NP_001171953.1 | 3 - 4 | 489 | 72 | |
| NM_013373.3 | NP_037505.1 | 3 - 4 | 489 | 72 | ||
| XM_006724239.2 | XP_006724302.1 | 3 - 4 | 477 | 72 | ||
| GenBank mRNA | AK131238.1 | - | 3 - 4 | 340 | 72 | |
| AY894890.1 | - | 3 - 4 | 489 | 72 | ||
| BC053544.1 | - | 3 - 4 | 489 | 5730191 | 72 | |
| CR456357.1 | - | 2 - 3 | 246 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000055.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22000711.hg.1 | ENST00000334554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000711.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000711.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000711.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000711.hg.1 | TC22000055.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000320602.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000468112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000469212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | ENST00000472497.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006643.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | NM_013373.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | TC2200006643.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000334554.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000405930.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | ENST00000436518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | NM_001185024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145603.hg.1 | NM_013373.3 | Refseq |

ZDHHC8
Genomic Map