Assay Details
| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198317.2 | NP_938073.1 | 2 - 3 | 476 | 84 | |
| XM_006710600.3 | XP_006710663.1 | 2 - 3 | 960 | 84 | ||
| XM_006710601.3 | XP_006710664.1 | 2 - 3 | 960 | 84 | ||
| GenBank mRNA | AY423763.1 | - | 2 - 3 | 476 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000021.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01000167.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000167.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000167.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000161.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000161.hg.1 | ENST00000455747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000161.hg.1 | ENST00000473277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000161.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000161.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000170.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000170.hg.1 | ENST00000455747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000170.hg.1 | ENST00000473277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000170.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000170.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000166.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000166.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000166.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000168.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000168.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000168.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000617073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | ENST00000455747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | ENST00000473277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | ENST00000540863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000162.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100144380.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144380.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144380.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144380.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144380.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144387.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000617073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144384.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144384.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144384.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144384.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144384.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144381.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144383.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144383.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144383.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144383.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144383.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144385.hg.1 | TC0100006479.hg.1 | Clariom D TC |

KLHL17
Genomic Map