Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182981.2 | NP_892026.1 | 2 - 3 | 249 | 75 | |
| GenBank mRNA | AK290383.1 | - | 2 - 3 | 231 | 75 | |
| AY037158.1 | - | 2 - 3 | 214 | 75 | ||
| AY258066.1 | - | 3 - 4 | 700 | 75 | ||
| AY258067.1 | - | 2 - 3 | 540 | 75 | ||
| AY258068.1 | - | 2 - 3 | 230 | 75 | ||
| BC020434.1 | - | 2 - 3 | 215 | 75 | ||
| BC032476.1 | - | 2 - 3 | 214 | 5215262 | 75 | |
| BC093687.1 | - | 2 - 3 | 132 | 7939532 | 75 | |
| BC113417.1 | - | 2 - 3 | 132 | 8322469 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000652.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009178.hg.1 | ENST00000343939.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009178.hg.1 | ENST00000361711.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009178.hg.1 | ENST00000393306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009178.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009178.hg.1 | TC16000652.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | TC1600011440.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191669.hg.1 | NM_182981.2 | Refseq |

OSGIN1
Genomic Map