Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011535315.2 | XP_011533617.1 | 1 - 2 | 239 | 121 | |
| XM_011535317.2 | XP_011533619.1 | 1 - 2 | 239 | 121 | ||
| XM_011535321.2 | XP_011533623.1 | 3 - 4 | 217 | 121 | ||
| XM_017020857.1 | XP_016876346.1 | 3 - 4 | 217 | 121 | ||
| XM_017020858.1 | XP_016876347.1 | 2 - 3 | 212 | 121 | ||
| XM_017020860.1 | XP_016876349.1 | 3 - 4 | 217 | 121 | ||
| XM_017020861.1 | XP_016876350.1 | 3 - 4 | 254 | 121 | ||
| XM_017020873.1 | XP_016876362.1 | 3 - 4 | 287 | 121 | ||
| GenBank mRNA | AK294343.1 | - | 1 - 2 | 47 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK294343.1, XM_011535315.2, XM_011535317.2, XM_011535321.2, XM_017020857.1, XM_017020858.1, XM_017020860.1, XM_017020861.1, XM_017020873.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000239852.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000379480.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000430347.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000452359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000473492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000480434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000492827.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000497679.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | ENST00000498245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008720.hg.1 | NM_004294.3 | Refseq |

MTRF1
Genomic Map