Assay Details
| Gene Symbol: KIAA1468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB040901.1 | - | 17 - 18 | 2192 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1468| Gene Symbol: KIAA1468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB040901.1 | - | 17 - 18 | 2192 | 142 | |
Set Membership: |
|