Assay Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000587.2 | NP_000578.2 | 14 - 15 | 1996 | 75 | |
| GenBank mRNA | AK290349.1 | - | 14 - 15 | 2006 | 75 | |
| AK312737.1 | - | 14 - 15 | 1930 | 75 | ||
| BC063851.1 | - | 14 - 15 | 1977 | 6184213 | 75 | |
| J03507.1 | - | 13 - 14 | 1882 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000187.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000489457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000489457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002195.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002195.hg.1 | ENST00000494960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002195.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002195.hg.1 | TC05000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002194.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002194.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002194.hg.1 | TC05000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002193.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002193.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002193.hg.1 | TC05000187.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000464864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000489457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000489457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000508185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007238.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000494960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148903.hg.1 | TC0500007238.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000313164.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000486779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | ENST00000513922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | NM_000587.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148902.hg.1 | TC0500007238.hg.1 | Clariom D TC |

C7
Genomic Map