Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014650.3 | NP_055465.1 | 1 - 2 | 426 | 94 | |
| GenBank mRNA | AK292827.1 | - | 1 - 2 | 159 | 94 | |
| BC002858.2 | - | 1 - 2 | 122 | 3636865 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001794.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292827.1,BC002858.2,NM_014650.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19024383.hg.1 | ENST00000221315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024383.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024383.hg.1 | TC19001794.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024382.hg.1 | ENST00000221315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024382.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024382.hg.1 | TC19001794.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024384.hg.1 | ENST00000221315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024384.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024384.hg.1 | TC19001794.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024381.hg.1 | ENST00000221315.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024381.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024381.hg.1 | TC19001794.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012020.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | TC1900012020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000594154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000598745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | ENST00000601310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207637.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | TC1900012020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | ENST00000597273.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207641.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207642.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207642.hg.1 | NM_014650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207642.hg.1 | TC1900012020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207642.hg.1 | ENST00000221315.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207642.hg.1 | NM_014650.3 | Refseq |

ZNF432
Genomic Map