| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024380.hg.1 |
ENST00000221315.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024380.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024380.hg.1 |
TC19001794.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
TC1900012020.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207628.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012020.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
NM_014650.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
TC1900012020.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000221315.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000594154.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000597273.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000598745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
ENST00000601310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207634.hg.1 |
NM_014650.3 |
Refseq |