Assay Details
| Gene Symbol: DPY19L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250895.3 | XP_005250952.1 | 2 - 3 | 257 | 142 | |
| XM_017013354.1 | XP_016868843.1 | 2 - 3 | 282 | 142 | ||
| XM_017013356.1 | XP_016868845.1 | 2 - 3 | 254 | 142 | ||
| XM_017013357.1 | XP_016868846.1 | 2 - 3 | 248 | 142 | ||
| GenBank mRNA | BP873374.1 | - | 2 - 3 | 251 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250895.3, XM_017013354.1, XM_017013356.1, XM_017013357.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000414645.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000519353.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000520774.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000521525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000522669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | ENST00000523020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008266.hg.1 | NM_181787.2 | Refseq |

DPY19L4
Genomic Map