| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025535.hg.1 |
ENST00000332836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025535.hg.1 |
ENST00000333452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025535.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19025535.hg.1 |
TC19001880.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
TC1900011494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197399.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000332836.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
ENST00000590200.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011494.hg.1 |
NM_176820.3 |
Refseq |