| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000974.hg.1 |
ENST00000380482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000974.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000974.hg.1 |
TC13000103.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000975.hg.1 |
ENST00000380482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000975.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000975.hg.1 |
TC13000103.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000978.hg.1 |
ENST00000380482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000978.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000978.hg.1 |
TC13000103.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000977.hg.1 |
ENST00000380482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000977.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000977.hg.1 |
TC13000103.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000976.hg.1 |
ENST00000380482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000976.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000976.hg.1 |
TC13000103.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300146016.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146016.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146016.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146016.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146016.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006774.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146014.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146014.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146014.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146014.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146014.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146015.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146015.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146015.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146015.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146015.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146018.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
ENST00000428944.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146019.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146017.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146017.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146017.hg.1 |
ENST00000380482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146017.hg.1 |
NM_032849.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146017.hg.1 |
TC1300006774.hg.1 |
Clariom D TC |