Assay Details
| Gene Symbol: APLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001642.2 | NP_001633.1 | 14 - 15 | 2036 | 74 | |
| NR_024516.1 | 11 - 12 | 1570 | 74 | |||
| GenBank mRNA | AF168956.1 | - | 14 - 15 | 1882 | 74 | |
| AK300378.1 | - | 11 - 12 | 1497 | 74 | ||
| L09209.1 | - | 14 - 15 | 1951 | 74 | ||
| L23114.1 | - | 12 - 13 | 1599 | 74 | ||
| L27631.1 | - | 14 - 15 | 1952 | 74 | ||
| S60099.1 | - | 14 - 15 | 1951 | 74 | ||
| Z22572.1 | - | 14 - 15 | 1946 | 74 | ||
More Information
Set Membership: |
|

APLP2
Genomic Map