| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006016.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006016.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006016.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006016.hg.1 |
TC0X000468.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153751.hg.1 |
TC0X00007918.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |