| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008080.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008080.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008080.hg.1 |
TC10000697.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008082.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008082.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008082.hg.1 |
TC10000697.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008081.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008081.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008081.hg.1 |
TC10000697.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008579.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158488.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158488.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158488.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158488.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158488.hg.1 |
TC1000008579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000158489.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158489.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158489.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158489.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158489.hg.1 |
TC1000008579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000158487.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158487.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158487.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158487.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158487.hg.1 |
TC1000008579.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000158486.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158486.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158486.hg.1 |
ENST00000370664.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000158486.hg.1 |
NM_024954.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000158486.hg.1 |
TC1000008579.hg.1 |
Clariom D TC |