| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018919.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018919.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018919.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018919.hg.1 |
TC10001497.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011368.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
ENST00000371930.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
ENST00000476963.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
NM_144590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177220.hg.1 |
TC1000011368.hg.1 |
Clariom D TC |