Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262186.2 | XP_005262243.1 | 3 - 4 | 740 | 88 | |
| XM_005262189.2 | XP_005262246.1 | 3 - 4 | 740 | 88 | ||
| XM_011531020.1 | XP_011529322.1 | 3 - 4 | 740 | 88 | ||
| GenBank mRNA | AK308097.1 | - | 3 - 4 | 430 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005262186.2, XM_005262189.2, XM_011531020.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | ENST00000584664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000276123.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000360700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000373165.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | ENST00000584664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007781.hg.1 | NM_021998.4 | Refseq |

ZNF711
Genomic Map