| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
ENST00000584664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
ENST00000276123.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
ENST00000373165.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
ENST00000542798.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005806.hg.1 |
TC0X000442.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
ENST00000584664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007781.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000276123.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000360700.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
ENST00000373165.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
NM_021998.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153177.hg.1 |
TC0X00007781.hg.1 |
Clariom D TC |