Assay Details
| Gene Symbol: AGPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012727.1 | NP_001012745.1 | 2 - 3 | 413 | 58 | |
| NM_006412.3 | NP_006403.2 | 2 - 3 | 413 | 58 | ||
| GenBank mRNA | AF000237.1 | - | 2 - 3 | 377 | 58 | |
| AF011374.1 | - | 2 - 3 | 371 | 58 | ||
| BC000026.2 | - | 2 - 3 | 356 | 3507150 | 58 | |
| BC004529.2 | - | 2 - 3 | 347 | 3347049 | 58 | |
| BC019292.1 | - | 2 - 3 | 365 | 2823580 | 58 | |
| BF347518.1 | - | 2 - 3 | 414 | 58 | ||
| CN407343.1 | - | 2 - 3 | 413 | 58 | ||
| U56418.2 | - | 2 - 3 | 350 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001738.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
epidermis development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to antidepressant
1-acylglycerol-3-phosphate O-acyltransferase activity
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
epidermis development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to antidepressant
1-acylglycerol-3-phosphate O-acyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | ENST00000371694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | ENST00000371696.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022313.hg.1 | TC09001738.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182232.hg.1 | TC0900011992.hg.1 | Clariom D TC |

AGPAT2
Genomic Map