Assay Details
| Gene Symbol: AGPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012727.1 | NP_001012745.1 | 4 - 5 | 674 | 165 | |
| NM_006412.3 | NP_006403.2 | 5 - 6 | 770 | 165 | ||
| GenBank mRNA | AF000237.1 | - | 5 - 6 | 734 | 165 | |
| AF011374.1 | - | 5 - 6 | 728 | 165 | ||
| BC000026.2 | - | 5 - 6 | 713 | 3507150 | 165 | |
| BC004529.2 | - | 4 - 5 | 608 | 3347049 | 165 | |
| BC007269.1 | - | 3 - 4 | 345 | 165 | ||
| BC019292.1 | - | 5 - 6 | 722 | 2823580 | 165 | |
| U56418.2 | - | 5 - 6 | 707 | 165 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001738.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
epidermis development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to antidepressant
1-acylglycerol-3-phosphate O-acyltransferase activity
positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
epidermis development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to antidepressant
1-acylglycerol-3-phosphate O-acyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | ENST00000371694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | ENST00000371696.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022309.hg.1 | TC09001738.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | ENST00000371694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | ENST00000371696.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022308.hg.1 | TC09001738.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000470861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011992.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182227.hg.1 | TC0900011992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000472820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182228.hg.1 | TC0900011992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000371694.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000371696.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | ENST00000538402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | NM_001012727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | NM_006412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182226.hg.1 | TC0900011992.hg.1 | Clariom D TC |

AGPAT2
Genomic Map