Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
ENST00000325215.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
ENST00000336783.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
ENST00000340394.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
ENST00000382686.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
ENST00000395547.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
NM_007245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
NM_145714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
NM_148414.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
NM_148415.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
NM_148416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004315.hg.1 |
TC16000308.hg.1 |
HTA TC |