Assay Details
| Gene Symbol: WNT8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300938.1 | NP_001287867.1 | 2 - 3 | 530 | 69 | |
| NM_001300939.1 | NP_001287868.1 | 2 - 3 | 530 | 69 | ||
| NM_058244.3 | NP_490645.1 | 3 - 4 | 438 | 69 | ||
| NR_125351.1 | 3 - 4 | 438 | 69 | |||
| XM_011543625.2 | XP_011541927.2 | 3 - 4 | 508 | 69 | ||
| XM_017009824.1 | XP_016865313.1 | 3 - 4 | 508 | 69 | ||
| XM_017009825.1 | XP_016865314.1 | 3 - 4 | 508 | 69 | ||
| XM_017009826.1 | XP_016865315.1 | 4 - 5 | 370 | 69 | ||
| GenBank mRNA | AB057725.1 | - | 3 - 4 | 245 | 69 | |
| AY009402.1 | - | 3 - 4 | 340 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017009826.1 transcripts.
Set Membership: |
|

WNT8A
Genomic Map