Assay Details
| Gene Symbol: MYH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005963.3 | NP_005954.3 | 38 - 39 | 5672 | 77 | |
| XM_017024675.1 | XP_016880164.1 | 38 - 39 | 5845 | 77 | ||
| GenBank mRNA | AJ573764.1 | - | 1 - 2 | 78 | 77 | |
| BC114545.1 | - | 36 - 37 | 5578 | 40070022 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001133.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17015372.hg.1 | ENST00000226207.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015372.hg.1 | ENST00000379814.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015372.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015372.hg.1 | TC17001133.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015370.hg.1 | ENST00000226207.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015370.hg.1 | ENST00000379814.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015370.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015370.hg.1 | TC17001133.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015371.hg.1 | ENST00000226207.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015371.hg.1 | ENST00000379814.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015371.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015371.hg.1 | TC17001133.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015373.hg.1 | ENST00000226207.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015373.hg.1 | ENST00000379814.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015373.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015373.hg.1 | TC17001133.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176143.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176143.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176143.hg.1 | TC1700009744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176143.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176143.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009744.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176141.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176141.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176141.hg.1 | TC1700009744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176141.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176141.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176140.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176140.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176140.hg.1 | TC1700009744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176140.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176140.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176142.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176142.hg.1 | NM_005963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176142.hg.1 | TC1700009744.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176142.hg.1 | ENST00000226207.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176142.hg.1 | NM_005963.3 | Refseq |

MYH1
Genomic Map