| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000358897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000375998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000375999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000401090.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000417793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
ENST00000471347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
NM_052929.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002992.hg.1 |
TC01000186.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000358897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000375998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000375999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000401090.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
ENST00000417793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
NM_052929.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002990.hg.1 |
TC01000186.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375996.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000375997.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000459961.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000472131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000483694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018175.hg.1 |
ENST00000524761.5 |
Ensembl |