| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
ENST00000358897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
ENST00000375998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
ENST00000375999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
ENST00000417793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
ENST00000471347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
NM_052929.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003002.hg.1 |
TC01000186.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000358897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000375995.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000375998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000375999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000401090.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000417793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
ENST00000471347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
NM_052929.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002994.hg.1 |
TC01000186.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
ENST00000358897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
ENST00000375998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
ENST00000375999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
ENST00000417793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
ENST00000471347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
NM_052929.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003003.hg.1 |
TC01000186.hg.1 |
HTA TC |