Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271816.1 | NP_001258745.1 | 2 - 3 | 1179 | 94 | |
| NM_014881.4 | NP_055696.3 | 1 - 2 | 1497 | 94 | ||
| XM_006718090.1 | XP_006718153.1 | 2 - 3 | 1191 | 94 | ||
| XM_011540429.1 | XP_011538731.1 | 2 - 3 | 1247 | 94 | ||
| GenBank mRNA | BC013124.2 | - | 1 - 2 | 1129 | 3048375 | 94 |
| BC062582.1 | - | 1 - 2 | 1129 | 5202821 | 94 | |
| D42045.1 | - | 1 - 2 | 1381 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001673.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via nonhomologous end joining
mitotic nuclear division
protection from non-homologous end joining at telomere
interstrand cross-link repair
cell division
nucleic acid phosphodiester bond hydrolysis
damaged DNA binding
5'-3' exodeoxyribonuclease activity
double-strand break repair via nonhomologous end joining
mitotic nuclear division
protection from non-homologous end joining at telomere
interstrand cross-link repair
cell division
nucleic acid phosphodiester bond hydrolysis
damaged DNA binding
5'-3' exodeoxyribonuclease activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10021950.hg.1 | ENST00000361384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021950.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021950.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021950.hg.1 | TC10001673.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021952.hg.1 | ENST00000361384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021952.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021952.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021952.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021952.hg.1 | TC10001673.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021954.hg.1 | ENST00000361384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021954.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021954.hg.1 | TC10001673.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021951.hg.1 | ENST00000361384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021951.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021951.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021951.hg.1 | TC10001673.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181350.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181356.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181356.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181356.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181356.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181356.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181348.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181351.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | ENST00000476112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011885.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | ENST00000369305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | NM_001271816.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181349.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181353.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181353.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181353.hg.1 | ENST00000361384.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181353.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181353.hg.1 | TC1000011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181354.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181354.hg.1 | NM_014881.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181354.hg.1 | TC1000011885.hg.1 | Clariom D TC |

DCLRE1A
Genomic Map