Assay Details
| Gene Symbol: PLXNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032242.3 | NP_115618.3 | 13 - 14 | 2884 | 99 | |
| XM_011512908.2 | XP_011511210.1 | 14 - 15 | 2986 | 99 | ||
| XM_011512909.1 | XP_011511211.1 | 14 - 15 | 3579 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000661.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
regulation of smooth muscle cell migration
branchiomotor neuron axon guidance
regulation of cell migration
regulation of axon extension involved in axon guidance
dichotomous subdivision of terminal units involved in salivary gland branching
semaphorin-plexin signaling pathway involved in axon guidance
neuron projection extension
receptor activity
semaphorin receptor activity
multicellular organism development
regulation of smooth muscle cell migration
branchiomotor neuron axon guidance
regulation of cell migration
regulation of axon extension involved in axon guidance
dichotomous subdivision of terminal units involved in salivary gland branching
semaphorin-plexin signaling pathway involved in axon guidance
neuron projection extension
receptor activity
semaphorin receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011870.hg.1 | ENST00000251772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011870.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011870.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011870.hg.1 | TC03000661.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300161693.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161693.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161693.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161693.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161693.hg.1 | TC0300008691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000393409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000503363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | ENST00000505278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008691.hg.1 | NM_032242.3 | Refseq |

PLXNA1
Genomic Map