Assay Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006417.4 | NP_006408.3 | 4 - 5 | 809 | 75 | |
| NR_135640.1 | 3 - 4 | 345 | 75 | |||
| NR_135641.1 | 4 - 5 | 812 | 75 | |||
| XM_005270380.3 | XP_005270437.1 | 4 - 5 | 2047 | 75 | ||
| XM_011540516.2 | XP_011538818.1 | 4 - 5 | 2047 | 75 | ||
| XM_017000083.1 | XP_016855572.1 | 4 - 5 | 2047 | 75 | ||
| GenBank mRNA | AK302527.1 | - | 4 - 5 | 770 | 75 | |
| AK316278.1 | - | 3 - 4 | 302 | 75 | ||
| AK316476.1 | - | 4 - 5 | 767 | 75 | ||
| BC022870.1 | - | 4 - 5 | 774 | 4071404 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000795.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK316278.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012720.hg.1 | ENST00000370747.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012720.hg.1 | ENST00000495254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012720.hg.1 | ENST00000545124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012720.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012720.hg.1 | TC01000795.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162942.hg.1 | TC0100008816.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |

IFI44
Genomic Map