Assay Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006417.4 | NP_006408.3 | 6 - 7 | 1136 | 139 | |
| XM_011540516.2 | XP_011538818.1 | 6 - 7 | 2374 | 139 | ||
| XM_017000083.1 | XP_016855572.1 | 6 - 7 | 2374 | 139 | ||
| GenBank mRNA | AK302527.1 | - | 6 - 7 | 1097 | 139 | |
| AK316278.1 | - | 5 - 6 | 629 | 139 | ||
| AK316476.1 | - | 6 - 7 | 1094 | 139 | ||
| BC022870.1 | - | 6 - 7 | 1101 | 4071404 | 139 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000795.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012724.hg.1 | ENST00000370747.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012724.hg.1 | ENST00000545124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012724.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012724.hg.1 | TC01000795.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012723.hg.1 | ENST00000370747.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012723.hg.1 | ENST00000495254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012723.hg.1 | ENST00000545124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012723.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01012723.hg.1 | TC01000795.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000438486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000446486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000467790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000476911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008816.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000472152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | ENST00000495254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162945.hg.1 | TC0100008816.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000370747.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000470323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | ENST00000485662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | NM_006417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100162946.hg.1 | TC0100008816.hg.1 | Clariom D TC |

IFI44
Genomic Map