Assay Details
| Gene Symbol: WNK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321299.1 | NP_001308228.1 | 13 - 14 | 2937 | 107 | |
| NM_032387.4 | NP_115763.2 | 14 - 15 | 3026 | 107 | ||
| XM_011525135.2 | XP_011523437.1 | 14 - 15 | 3956 | 107 | ||
| XM_017024962.1 | XP_016880451.1 | 14 - 15 | 3956 | 107 | ||
| XM_017024963.1 | XP_016880452.1 | 14 - 15 | 3953 | 107 | ||
| XM_017024964.1 | XP_016880453.1 | 13 - 14 | 3818 | 107 | ||
| XM_017024965.1 | XP_016880454.1 | 13 - 14 | 3815 | 107 | ||
| XM_017024966.1 | XP_016880455.1 | 11 - 12 | 2078 | 107 | ||
| GenBank mRNA | AF390018.1 | - | 14 - 15 | 2979 | 107 | |
| AJ316534.1 | - | 14 - 15 | 2975 | 107 | ||
| AK096003.1 | - | 8 - 9 | 1449 | 107 | ||
| BC136664.1 | - | 14 - 15 | 2976 | 9020654 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000536.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
ion transport
chloride transport
protein localization
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
regulation of cellular process
ion homeostasis
renal sodium ion absorption
distal tubule morphogenesis
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
protein serine/threonine kinase activity
protein binding
ATP binding
chloride channel inhibitor activity
protein phosphorylation
ion transport
chloride transport
protein localization
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
regulation of cellular process
ion homeostasis
renal sodium ion absorption
distal tubule morphogenesis
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
protein serine/threonine kinase activity
protein binding
ATP binding
chloride channel inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006529.hg.1 | ENST00000246914.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006529.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006529.hg.1 | TC17000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006526.hg.1 | ENST00000246914.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006526.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006526.hg.1 | TC17000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006530.hg.1 | ENST00000246914.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006530.hg.1 | ENST00000316085.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006530.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006530.hg.1 | TC17000536.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | TC1700007915.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157738.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | TC1700007915.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157736.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | TC1700007915.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157737.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000246914.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000587745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000591448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | ENST00000592669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007915.hg.1 | NM_032387.4 | Refseq |

WNK4
Genomic Map